missing translation for 'onlineSavingsMsg'
Learn More

L-glutamine, 99 to 100%, MP Biomedicals™

Product Code. 17827496 Shop All MP Biomedical Products
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
This item is not returnable. View return policy

Product Code. 17827496

Brand: MP Biomedicals 0210180683

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Color White
Assay Percent Range 99 to 100%
Quantity 250 g
Formula Weight 146.1
Physical Form Powder
Chemical Name or Material L-Glutamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.