Learn More
L-Glucono-1,5-lactone, TRC
Product Code. 30240878
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240878 100 mg 100mg
30240906 500 mg 500mg
30240633 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30240878 Supplier Toronto Research Chemicals Supplier No. TRCG413305100MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 52153-09-0
Molecular Formula C6 H10 O6
Molecular Weight (g/mol) 178.14
Synonym L-Gluconic acid, δ-lactone, (3S,4R,5R,6S)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, L-Glucono-1,5-lactone, L-Glucono-δ-lactone
IUPAC Name (3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
SMILES OC[C@@H]1OC(=O)[C@@H](O)[C@H](O)[C@H]1O
Spezifikation
Quantity 100 mg
InChI Formula InChI=1S/C6H10O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-5,7-10H,1H2/t2-,3-,4+,5-/m0/s1
Formula Weight 178.0477
Chemical Name or Material L-Glucono-1,5-lactone