Learn More
L-Dihydroorotic Acid-D3, TRC
Specifications
Specifications
| Chemical Name or Material | L-Dihydroorotic Acid-D3 |
| CAS | 300577-17-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C5 D3 H3 N2 O4 |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C5H6N2O4/c8-3-1-2(4(9)10)6-5(11)7-3/h2H,1H2,(H,9,10)(H2,6,7,8,11)/t2-/m0/s1/i1D2,2D |
| SMILES | [2H]C1([2H])C(=O)NC(=O)N[C@]1([2H])C(=O)O |
| IUPAC Name | (4S)-4,5,5-trideuterio-2,6-dioxo-1,3-diazinane-4-carboxylic acid |
| Molecular Weight (g/mol) | 161.131 |
| Formula Weight | 161.052 |
| Show More |