Learn More
L-Cysteine Sulfone, TRC
Product Code. 30136153
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136153 500 mg 500mg
30003211 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30136153 Supplier Toronto Research Chemicals Supplier No. TRCC997825500MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30452-69-8
Molecular Formula C6 H12 N2 O6 S2
Molecular Weight (g/mol) 272.3
IUPAC Name (2R)-2-amino-3-[(2R)-2-amino-2-carboxyethyl]sulfonylsulfanylpropanoic acid
SMILES N[C@@H](CSS(=O)(=O)C[C@H](N)C(=O)O)C(=O)O
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C6H12N2O6S2/c7-3(5(9)10)1-15-16(13,14)2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1
Formula Weight 272.0137
Chemical Name or Material L-Cysteine-sulfone