Learn More
L-Cysteic acid monohydrate, 99%, Thermo Scientific Chemicals
Product Code. 15216038 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 15216038
Brand: Thermo Scientific Acros 452840100
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 23537-25-9
Molecular Formula C3H6NO5S
Molecular Weight (g/mol) 168.14
MDL Number MFCD00149544
InChI Key XVOYSCVBGLVSOL-REOHCLBHSA-M
Synonym l-cysteic acid monohydrate, unii-vn8x20t18r, cysteic acid monohydrate, l, r-2-amino-3-sulfopropanoic acid hydrate, l-alanine, 3-sulfo-, monohydrate, cysteinesulfonic acid hydrate, h-cys o 2-oh.h2o, l-cysteic acid hydrate, l-cysteic acid monohydrate mi, alanine, 3-sulfo-, monohydrate, l
PubChem CID 12308854
IUPAC Name (2R)-2-amino-3-sulfopropanoic acid;hydrate
SMILES [NH3+][C@@H](CS([O-])(=O)=O)C([O-])=O
Specifications
Melting Point 253.0°C to 254.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Linear Formula HO3SCH2CH(NH2)CO2H·H2O
Quantity 10 g
Specific Rotation Condition + 8.00 (20.00°C c=5, H2O)
Specific Rotation + 8.00
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 187.17
Percent Purity 99%
Chemical Name or Material L-Cysteic acid monohydrate