Learn More
L-Citrulline, TRC
Product Code. 30136113
Change view
Click to view available options
Quantity:
10 mg
1 g
5 g
Unit Size:
10mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136113 5 g 5g
30009770 10 mg 10mg
30012235 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136113 Supplier Toronto Research Chemicals Supplier No. TRCC5357005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 372-75-8
Molecular Formula C6 H13 N3 O3
Molecular Weight (g/mol) 175.19
Synonym N5-(Aminocarbonyl)-L-ornithine, Ornithine, N5-carbamoyl-, L- (8CI), Cit, Citrulline, L-(+)-Citrulline, L-Citrulline, N5-Carbamoyl-L-ornithine, NSC 27425, Nδ-Carbamylornithine, α-Amino-δ-ureidovaleric acid, δ-Ureidonorvaline
IUPAC Name (2S)-2-amino-5-(carbamoylamino)pentanoic acid
SMILES N[C@@H](CCCNC(=O)N)C(=O)O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C6H13N3O3/c7-4(5(10)11)2-1-3-9-6(8)12/h4H,1-3,7H2,(H,10,11)(H3,8,9,12)/t4-/m0/s1
Formula Weight 175.0957
Chemical Name or Material L-Citrulline