Learn More
L-Citrulline, 99.3%, MP Biomedicals™
Product Code. 11413350 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413350 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11413350 Supplier MP Biomedicals Supplier No. 0210139780
Chemical Identifiers
CAS 372-75-8
Molecular Formula C6H13N3O3
Molecular Weight (g/mol) 175.19
MDL Number MFCD00064397
InChI Key RHGKLRLOHDJJDR-UHFFFAOYNA-N
Synonym l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
PubChem CID 9750
ChEBI CHEBI:16349
SMILES NC(CCCNC(N)=O)C(O)=O
Specifications
Color White
Assay Percent Range 99.3%
IUPAC Name (2S)-2-amino-5-(carbamoylamino)pentanoic acid
Quantity 100 g
Formula Weight 175.19
Physical Form Crystalline Powder
Chemical Name or Material L-Citrulline