Learn More
L-Aspartic Acid Dibenzyl Ester p-Toluenesulfonate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | L-Aspartic Acid Dibenzyl Ester p-Toluenesulfonate Salt |
| CAS | 2886-33-1 |
| Molecular Formula | C25H27O7S |
| Quantity | 10 g |
| Synonym | L-Aspartic Acid 1,4-bis(Phenylmethyl) Ester 4-Methylbenzenesulfonate, Aspartic Acid Dibenzyl Ester p-Toluenesulfonate, L-Aspartic Acid bis(Phenylmethyl) Ester 4-Methylbenzenesulfonate, Dibenzyl L-Aspartate Tosylate, Dibenzyl Aspartate p-Tosylate, Dibenzyl Aspartate Tosylate, Dibenzyl Aspartate Tosylate Salt, L-Aspartic Acid Dibenzyl Ester 4-Toluenesulfonate Salt, L-Aspartic Acid Dibenzyl Ester p-toluenesulfonate, L-Aspartic Acid Dibenzyl Ester p-Toluenesulfonate Salt, L-Aspartic Acid Dibenzyl Ester Tosylate, L-Aspartic Acid alpha,alpha-Dibenzyl Ester Tosylate, S-Aspartic Acid Dibenzyl Ester p-Tosylate |
| InChI Formula | InChI=1S/C18H19NO4.C7H8O3S/c19-16(18(21)23-13-15-9-5-2-6-10-15)11-17(20)22-12-14-7-3-1-4-8-14;1-6-2-4-7(5-3-6)11(8,9)10/h1-10,16H,11-13,19H2;2-5H,1H3,(H,8,9,10)/t16-;/m0./s1 |
| SMILES | Cc1ccc(cc1)S(=O)(=O)O.N[C@@H](CC(=O)OCc2ccccc2)C(=O)OCc3ccccc3 |
| IUPAC Name | dibenzyl (2S)-2-aminobutanedioate;4-methylbenzenesulfonic acid |
| Molecular Weight (g/mol) | 485.55 |
| Formula Weight | 485.1508 |