Learn More
L-Arginine-2,3,3,4,4,5,5-d7 HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | L-Arginine d7 hydrochloride |
| CAS | 204244-77-9 |
| Molecular Formula | C6 D7 H7 N4 O2 . H Cl |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1 H/t4-;/m0./s1/i1D2,2D2,3D2,4 D; |
| SMILES | Cl.[2 H]C([2 H])(NC(=N)N)C([2 H])([2 H])C([2 H])([2 H])[C@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-2,3,3,4,4,5,5-heptadeuterio-5-guanidino-pentanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 217.7 |
| Formula Weight | 217.1323 g/mol |
| Percent Purity | 98 atom % D, min 97% Chemical Purity |