missing translation for 'onlineSavingsMsg'
Learn More

L-Arabinitol, TRC

Product Code. 17948654
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17948654 10 g 10g
17939964 2.5 g 2.5g
17929824 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17948654 Supplier Toronto Research Chemicals Supplier No. TRCA73561010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7643-75-6
Molecular Formula C5 H12 O5
Molecular Weight (g/mol) 152.15
Synonym Arabinitol, L- (8CI), (-)-Arabitol, L-(-)-Arabitol, L-Arabitol, L-Lyxitol, L-arabino-Pentitol, NSC 83253
IUPAC Name (2S,4S)-pentane-1,2,3,4,5-pentol
SMILES OC[C@H](O)C(O)[C@@H](O)CO

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4-/m0/s1
Formula Weight 152.0685
Chemical Name or Material L-(-)-Arabitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.