missing translation for 'onlineSavingsMsg'
Learn More

L-Alpha-Acetoxypropionyl Chloride, TRC

Product Code. 30314667
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314667 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314667 Supplier Toronto Research Chemicals Supplier No. TRCA1677431G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-alpha-Acetoxypropionyl Chloride
CAS 36394-75-9
Molecular Formula C5H7ClO3
Quantity 1 g
Synonym (2S)-2-(Acetyloxy)propanoyl Chloride, (1S)-2-Chloro-1-methyl-2-oxoethyl Acetate, (2S)-1-Chloro-1-oxopropan-2-yl Acetate, (2S)-2-Acetoxypropionyl Chloride, (S)-(-)-2-Acetoxypropionyl Chloride, (S)-1-Chloro-1-oxopropan-2-yl Acetate, (S)-2-(Acetyloxy)propanoyl Chloride, (S)-2-Acetoxypropanoyl Chloride, (S)-2-Acetoxypropionoyl Chloride, (S)-2-Acetoxypropionyl Chloride, (S)-2-Acetyloxypropionyl Chloride, 2-(S)-Acetoxypropanoyl Chloride, Acetic Acid (1S)-1-(Chlorocarbonyl)ethyl Ester, Acetic Acid (1S)-2-Chloro-1-methyl-2-oxoethyl Ester, Acetic Acid (2S)-1-Chloro-1-oxopropan-2-yl Ester
InChI Formula InChI=1S/C5H7ClO3/c1-3(5(6)8)9-4(2)7/h3H,1-2H3/t3-/m0/s1
SMILES C[C@H](OC(=O)C)C(=O)Cl
IUPAC Name [(2S)-1-chloro-1-oxopropan-2-yl] acetate
Molecular Weight (g/mol) 150.56
Formula Weight 150.0084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.