missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine Benzyl Ester p-Toluenesulfonate Salt, TRC

Product Code. 30321441
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321441 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321441 Supplier Toronto Research Chemicals Supplier No. TRCA48151510G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Alanine Benzyl Ester p-Toluenesulfonate Salt
CAS 42854-62-6
Molecular Formula C10 H13 N O2 . C7 H8 O3 S
Quantity 10 g
InChI Formula InChI=1S/C10H13NO2.C7H8O3S/c1-8(11)10(12)13-7-9-5-3-2-4-6-9;1-6-2-4-7(5-3-6)11(8,9)10/h2-6,8H,7,11H2,1H3;2-5H,1H3,(H,8,9,10)/t8-;/m0./s1
SMILES C[C@H](N)C(=O)OCc1ccccc1.Cc2ccc(cc2)S(=O)(=O)O
IUPAC Name benzyl (2S)-2-aminopropanoate;4-methylbenzenesulfonic acid
Molecular Weight (g/mol) 351.42
Formula Weight 351.114
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.