missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine benzyl ester p-toluenesulfonate, 98%, Thermo Scientific™

Product Code. p-7058354
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332976 1 g 1g
11342976 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332976 Supplier Thermo Scientific Alfa Aesar Supplier No. L10743.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42854-62-6
Molecular Formula C17H21NO5S
Molecular Weight (g/mol) 351.42
MDL Number MFCD00066143,MFCD00066142
InChI Key NWOPHJSSBMABBD-UHFFFAOYNA-N
Synonym l-alanine benzyl ester 4-toluenesulfonate, l-alanine benzyl ester p-toluenesulfonate, ala-obzl tosoh, l-alanine benzyl ester p-toluenesulfonate salt, l-alanine benzyl ester tosylate, h-ala-obzl.tosoh, s-benzyl 2-aminopropanoate 4-methylbenzenesulfonate, benzyl 2s-2-aminopropanoate 4-toluenesulfonate, l-ala-obel.tsoh, benzyl 2s-2-aminopropanoate; para-toluene sulfonate
PubChem CID 2802424
IUPAC Name benzyl (2S)-2-aminopropanoate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.CC(N)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 114°C to 116°C
Beilstein 3586474
Quantity 1 g
Formula Weight 351.42
Optical Rotation −5.3° (c=3 in Methanol)
Percent Purity 98%
Chemical Name or Material L-Alanine benzyl ester p-toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.