Learn More
L-Alanine benzyl ester p-toluenesulfonate, 98%, Thermo Scientific™
Product Code. 11342976
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342976 5 g 5g
11332976 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11342976 Supplier Thermo Scientific Alfa Aesar Supplier No. L10743.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42854-62-6
Molecular Formula C17H21NO5S
Molecular Weight (g/mol) 351.42
MDL Number MFCD00066143,MFCD00066142
InChI Key NWOPHJSSBMABBD-UHFFFAOYNA-N
Synonym l-alanine benzyl ester 4-toluenesulfonate, l-alanine benzyl ester p-toluenesulfonate, ala-obzl tosoh, l-alanine benzyl ester p-toluenesulfonate salt, l-alanine benzyl ester tosylate, h-ala-obzl.tosoh, s-benzyl 2-aminopropanoate 4-methylbenzenesulfonate, benzyl 2s-2-aminopropanoate 4-toluenesulfonate, l-ala-obel.tsoh, benzyl 2s-2-aminopropanoate; para-toluene sulfonate
PubChem CID 2802424
IUPAC Name benzyl (2S)-2-aminopropanoate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.CC(N)C(=O)OCC1=CC=CC=C1
Specifications
Melting Point 114°C to 116°C
Beilstein 3586474
Quantity 5 g
Formula Weight 351.42
Optical Rotation −5.3° (c=3 in Methanol)
Percent Purity 98%
Chemical Name or Material L-Alanine benzyl ester p-toluenesulfonate