Learn More
L-Alanine-13C3, TRC
Product Code. 17959274
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17959274 5 mg 5mg
17908904 1 mg 1mg
17928914 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17959274 Supplier Toronto Research Chemicals Supplier No. TRCA4815045MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100108-77-8
Molecular Formula 13C3H7NO2
Molecular Weight (g/mol) 92.07
Synonym L-Alanine-1,2,3-13C3
IUPAC Name (2S)-2-amino(1,2,3-13C3)propanoic acid
SMILES O[13C]([13C@@H](N)[13CH3])=O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1/i1+1,2+1,3+1
Percent Purity >95%
Chemical Name or Material L-Alanine-13C3
Grade HPLC