missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-741,626
GREENER_CHOICE

Product Code. 30001160
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001160

Brand: Tocris Bioscience™ 1003/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity D2 antagonist

A potent D2 dopamine receptor selective antagonist, with affinities of 2.4, 100 and 220 nM for D2, D3 and D4 receptors respectively. Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81226-60-0
Molecular Formula C20H21ClN2O

Specifications

Quantity 10 mg
Formula Weight 340.85
Purity >98%
Molecular Weight (g/mol) 340.851
Chemical Name or Material L-741, 626
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.