missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-701,324
GREENER_CHOICE

Product Code. 18025592
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18025592

Brand: Tocris Bioscience™ 0907/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NMDA antagonist; acts at glycine site

An orally active and long acting anticonvulsant with high affinity and selectivity for the glycine site on the NMDA receptor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142326-59-8
Molecular Formula C21H14ClNO3

Specifications

Quantity 10 mg
Formula Weight 363.8
Purity >99%
Molecular Weight (g/mol) 363.797
Chemical Name or Material L-701, 324
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.