missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L-701,324

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
An orally active and long acting anticonvulsant with high affinity and selectivity for the glycine site on the NMDA receptor.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 363.8 |
| Purity | >99% |
| Molecular Weight (g/mol) | 363.797 |
| Chemical Name or Material | L-701, 324 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction