missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ L-685,458

Product Code. 18045312
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18045312 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18045312 Supplier R&D Systems™ Supplier No. 2627/1
  • ONLINE EXCLUSIVE PRICE
    430.00€
    406.35€ / 1mg
    Save 23.65€
    5% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective γ-secretase inhibitor

Potent and selective γ-secretase inhibitor (IC50 = 17 nM) that displays > 50-fold selectivity over a range of aspartyl, serine and cysteine proteases. Exhibits equal potency for inhibition of Aβ40 and Aβ42 peptides (IC50 values are 48 and 67 nM respectively in human neuroblastoma cells). Also regulates CXCR4 and VEGFR2 expression through inhibition of Notch signaling in vitro.
TRUSTED_SUSTAINABILITY

Specifications

CAS 292632-98-5
Quantity 1 mg
Product Line Tocris
Molecular Formula C39H52N4O6
Formula Weight 672.85
Purity >97%
Molecular Weight (g/mol) 672.867
Chemical Name or Material L-685, 458
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.