missing translation for 'onlineSavingsMsg'
Learn More

L-5-Hydroxytryptophan hydrate, 98%, Thermo Scientific Chemicals

Product Code. 11461698
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11461698

Brand: Thermo Scientific Alfa Aesar A13954.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9-8-4350
MDL Number MFCD00064341
ChEBI CHEBI:17780

Specifications

Melting Point 258°C (decomposition)
Molecular Formula C11H12N2O3
UN Number UN2811
Beilstein 88200
Merck Index 14,4847
Synonym 5-hydroxy-l-tryptophan, l-5-hydroxytryptophan, oxitriptan, 5-hydroxytryptophan, levothym, tript-oh, cincofarm, l-5-htp, pretonine, 2s-2-amino-3-5-hydroxy-1h-indol-3-yl propanoic acid
InChI Key LDCYZAJDBXYCGN-IMWMWJONNA-N
SMILES N[C@@H](CC1=CNC2=CC=C(O)C=C12)C(O)=O
IUPAC Name (2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 220.23
Quantity 5 g
PubChem CID 439280
Formula Weight 220.23 (Anhydrous)
Optical Rotation -33°C=1 aq. soln.)
Percent Purity 98%
Chemical Name or Material L-5-Hydroxytryptophan hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.