missing translation for 'onlineSavingsMsg'
Learn More

L-(2S,3aS,7aS)-Octahydroindole-2-carboxylic Acid, TRC

Product Code. 30420123
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420123 1 g -
30414347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30420123 Supplier Toronto Research Chemicals Supplier No. TRCO2369001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80875-98-5
Molecular Formula C9 H15 N O2
Molecular Weight (g/mol) 169.22
Synonym 1H-Indole-2-carboxylic acid, octahydro-, [2S-(2alpha,3beta,7beta)]-, (2S,3AS,7aS)-2-carboxyoctahydroindole, (2S,3AS,7aS)-Octahydroindole-2-carboxylic acid, (2S,3aS,7aS)-perhydroindole-2-carboxylic acid, Perindopril tert-Butylamine Imp. A (EP)
IUPAC Name (2S,3aS,7aS)-2,3,3a,4,5,6,7,7a-octahydro-1H-indole-2-carboxylic acid
SMILES OC(=O)[C@@H]1C[C@@H]2CCCC[C@@H]2N1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H15NO2/c11-9(12)8-5-6-3-1-2-4-7(6)10-8/h6-8,10H,1-5H2,(H,11,12)/t6-,7-,8-/m0/s1
Formula Weight 169.1103
Percent Purity >95%
Chemical Name or Material (2S,3aS,7aS)-octahydro-1Hindole-2-carboxylic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.