missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L-168,049

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Very potent and selective, non-competitive antagonist of the human glucagon receptor (hGR). Binds with high affinity to human GR (IC50 = 3.7 nM), and moderate affinity to murine and canine GRs (IC50 values are 63 and 60 nM respectively). In contrast, displays poor affinity for rat, guinea pig, and rabbit glucagon receptors (IC50 > 1 μM). In functional studies, inhibits glucagon-stimulated cAMP synthesis in CHO cells expressing hGR (IC50 = 41 nM), and in murine liver membranes. Orally active in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 467.79 |
| Purity | >99% |
| Molecular Weight (g/mol) | 467.791 |
| Chemical Name or Material | L-168, 049 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction