Learn More
Kynurenic-3,5,6,7,8-d5 Acid, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Kynurenic Acid-D5 |
| CAS | 350820-13-2 |
| Molecular Formula | C10 2H5 H2 N O3 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C10H7NO3/c12-9-5-8(10(13)14)11-7-4-2-1-3-6(7)9/h1-5 H,(H,11,12)(H,13,14)/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(O)c([2 H])c(nc2c1[2 H])C(=O)O |
| IUPAC Name | 3,5,6,7,8-pentadeuterio-4-hydroxyquinoline-2-carboxylic acid |
| Molecular Weight (g/mol) | 194.2 |
| Formula Weight | 194.074 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |