Learn More
IKA™ KS 501 Digital Orbital Shaker
Product Code. 15628155 Shop All IKA Products
Change view
Click to view available options
Plug Type:
EU Plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
15628155 EU Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15628155 Supplier IKA™ Supplier No. 0025002985
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Low-profile laboratory orbital shaker with large mounting surface.
  • Orbital diameter 30 mm
  • Ideal for vessels with a volume of more than 250 mL, e.g., round flasks, Erlenmeyer flasks, culture flasks and bottles
  • Ideal for biological and microbiological growth tests and for production of stock solutions
  • Continuous operation (endless) even under extreme loads
  • Timer function
  • Attachments are not included, please order separately
  • Integrated USB and RS 232 interface
TRUSTED_SUSTAINABILITY
Specifications
Load Bearing Capacity (Metric) 15 kg
Speed Range 0 to 300 rpm
Electrical Requirements 230/115/100 V, 50/60 Hz, 74 W
Voltage 100/115/230 V
Wattage 74 W
Plug Type EU Plug
Frequency 50/60 Hz
Dimensions (L x W x H) 505 x 120 x 585 mm
Weight (Metric) 26 kg
Model KS 501 Digital
Display 7 Segment LED
Includes Integrated USB and RS 232 interface, EU Plug