Learn More
Novus Biologicals™ KRT28 293T Cell Transient Overexpression Lysate
Product Code. 18555191 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18555191 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18555191 Supplier Novus Biologicals™ Supplier No. H00162605T01
 Restricted Item
Estimated Shipment: 17-08-2026
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target KRT28
Synonym cytokeratin-28, K25D, K28, keratin 25 D, keratin 28, keratin, type I cytoskeletal 28, keratin-28, KRT25D
Gene Symbol KRT28