Learn More
Thermo Scientific Chemicals Krebs-Ringer Solution, bicarbonate-buffered
Product Code. 15428769
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428769 1 L 1 L
15418769 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15428769 Supplier Thermo Scientific Chemicals Supplier No. J67591.K2
 Restricted Item
In Stock
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Physical Form Liquid
Quantity 1 L
Packaging Plastic Bottle
What is the pH of the Krebs-Ringer Solution, bicarbonate-buffered?
The pH of the Krebs-Ringer Solution, bicarbonate-buffered is 7.3.
Is the Krebs-Ringer Solution, bicarbonate-buffered sterile?
The Krebs-Ringer Solution, bicarbonate-buffered is prepared in 18.2 megohms water and filtered through 0.22 micron filter to remove contaminants. No sterility testing is performed so it cannot be certified as sterile.
What are the concentrations of the components of Krebs-Ringer Solution, bicarbonate-buffered?
The Krebs-Ringer Solution, bicarbonate-buffered contains 120 mM sodium chloride, 5 mM potassium chloride, 2 mM calcium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5 mM D-glucose.