missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KRAS (Human) Recombinant Protein
Human KRAS full-length ORF ( AAH13572, 1 a.a. - 188 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003845-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MTEYKLVVVGAGGVGKSALTIQLIQNHFVDEYDPTIEDSYRKQVVIDGETCLLDILDTAGHEEYSAMRDQYMRTGEGFLCVFAINNTKSFEDIHHYREQIKRVKDSEDVPMVLVGNKCDLPSRTVDTKQAQDLARSYGIPFIETSAKTRQGVDDAFYTLVREIRKHKEKMSKDGKKKKKKSKTKCVIM
Specifications
AAH13572 | |
v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KRAS | |
GST |
3845 | |
Wheat germ expression system | |
25 μg | |
C-K-RAS, K-RAS2A, K-RAS2B, K-RAS4A, K-RAS4B, KI-RAS, KRAS1, KRAS2, NS3, RASK2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |