missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KPNA5 (Human) Recombinant Protein
Human KPNA5 full-length ORF ( AAH47409.1, 1 a.a. - 539 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003841-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDAMASPGKDNYRMKSYKNKALNPQEMRRRREEEGIQLRKQKREEQLFKRRNVYLPRNDESMLESPIQDPDISSTVPIPEEEVVTTDMVQMIFSNNADQQLTATQKFRKLLSKEPNPPIDQVIQKPGVVQRFVKFLERNENCTLQFEAAWALTNIASGTFLHTKVVIETGAVPIFIKLLNSEHEDVQEQAVWALGNIAGDNAECRDFVLNCEILPPLLELLTNSNRLTTTRNAVWALSNLCRGKNPPPNFSKVSPCLNVLSRLLFSSDPDVLADVCWALSYLSDGPNDKIQAVIDSGVCRRLVELLMHNDYKVVSPALRAVGNIVTGDDIQTQVILNCSALPCLLHLLSSPKESIRKEACWTVSNITAGNRAQIQAVIDANIFPVLIEILQKAEFRTRKEAAWAITNATSGGTPEQIRYLVALGCIKPLCDLLTVMDSKIVQVALNGLENILRLGEQESKQNGIGINPYCALIEEAYGLDKIEFLQSHENQEIYQKAFDLIEHYFGVEEDDPSIVPQVDENQQQFIFQQQEAPMDGFQL
Specifications
AAH47409.1 | |
karyopherin alpha 5 (importin alpha 6) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KPNA5 | |
GST |
3841 | |
Wheat germ expression system | |
10 μg | |
IPOA6, SRP6 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |