Learn More
DWK Life Sciences Kontes™ Ultra-Ware™ Microfiltration Assembly with Stainless Steel Support and Ground Joint (47mm)
Product Code. 11898512
Change view
Click to view available options
Microfiltration Assembly, 47mm Filter Holders With Flasks, Funnel: 300mL, Support Material: 316 SS
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11898512 Microfiltration Assembly, 47mm Filter Holders With Flasks, Funnel: 300mL, Support Material: 316 SS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11898512 Supplier DWK Life Sciences Supplier No. 9538551047
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Recommended when filtering viscous or proteinaceous solutions to give the maximum flow rate
  • This apparatus is also used to produce ultra-clean filtrate since the stainless steel screen will not shed particles into the filtrate
  • The ground joint connection eliminates the possible phthalate contamination that can occur when using silicone or neoprene stoppers
  • Each apparatus is supplied with a funnel, an anodized aluminum clamp, a stainless steel support screen, a PTFE gasket, a 47mm glass support base and a filtration flask
  • 80 x 80 304 stainless steel mesh, 0.0055 in. opening size, 19.4% open area, 0.007 in. wire diameter
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (316)
Type Microfiltration Assembly
Volume (Metric) 1 L
Width (English) 5.37 in.
Width (Metric) 11.7 cm
Height (English) 17 in.
Height (Metric) 43 cm