Learn More
DWK Life Sciences KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
Product Code. 11868522
Change view
Click to view available options
Volume (Metric):
1000 mL (Funnel), 4000 mL (Flask)
300 mL (Funnel), 1000 mL (Flask)
500 mL (Funnel), 2000 mL (Flask)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11868522 500 mL (Funnel), 2000 mL (Flask) Each
11858482 300 mL (Funnel), 1000 mL (Flask) Each
11719055 1000 mL (Funnel), 4000 mL (Flask) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11868522 Supplier DWK Life Sciences Supplier No. 9538350000
  • ONLINE EXCLUSIVE PRICE
    865.00€
    810.79€ / Pack of 1
    Save 54.21€
    6% Off
 Restricted Item
Estimated Shipment: 08-07-2027
KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm), for routine filtration of corrosive liquids and removal of particulates from HPLC solvents.
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
  • Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
  • Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
  • Support base has a 40-60 micron porosity glass frit and an integral vacuum connection located above the drip tip to prevent contamination of the vacuum line with filtrate droplets
  • Each apparatus is supplied with a funnel, an anodized aluminum clamp, a 47mm fritted glass support base and a filtration flask
  • Joints are 40/35
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Disposable Reusable
Material Glass
Sterility Non-sterile
Type Microfiltration Assembly
Volume (Metric) 500 mL (Funnel), 2000 mL (Flask)
For Use With (Equipment) Filtration Assembly and vacuum pump