Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass support and GL 45 Style Bottle (47mm)
Product Code. 11832923
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11832923 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11832923 Supplier DWK Life Sciences Supplier No. 9537505347
Designed to collect filtrate directly into a media-style bottle
  • Supplied with funnel, clamp, fritted glass support base, tubing adapter, PBT cap, sealing ring and vacuum/pressure bottle
  • Support base connects directly to bottle with unique cap and sealing ring
  • Sealed-in 40-60 micron porosity fritted glass disc
  • Vacuum and pressure rated from -1 to 1.5 bar
  • Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly
  • All components are steam autoclavable
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Type Microfiltration Assembly
Volume (Metric) 1 L
Width (English) 4 in.
Width (Metric) 10.2 cm
Height (English) 14.5 in.
Height (Metric) 36.8 cm