Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support (25mm)
Product Code. 11849913
Change view
Click to view available options
Volume (Metric):
15 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11849913 15 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11849913 Supplier DWK Life Sciences Supplier No. 9537050000
Only null left
Add to basket
Add to basket
Designed to handle small volumes of liquids for analysis of particulate or microbiological contamination
  • Supplied with a 40-60 micron porosity fritted glass support base, a 15mL graduated funnel, an anodized aluminum clamp and a No. 5 silicone stopper
  • Funnels are graduated from 5 to 15mL in 1mL increments
  • Prefilter size is 16mm diameter
  • Approximate filter area is 2.5cm2
  • Connection to our 125mL filtration flask (available as an accessory) is made with a No. 5 silicone stopper
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Volume (Metric) 15 mL
Width (English) 1 in.
Width (Metric) 2.5 cm
Height (English) 6 in.
Height (Metric) 15.2 cm