Learn More
DWK Life Sciences Kimble™ Kontes™ Reaction-Jacketed Beakers
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
Description
The open top provides easy access to concentrated residual samples.
- A conical lower interior surface eliminates spin-out when used with an egg-shaped PTFE magnetic stir bar
- Jacket has serrated 3/8 in. hose connections
- Design meets ASTM Specification E960, Type I requirements
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Capacity (English) | 33.81 oz. |
| Description | Connection: 0.37 in. Serrated Hose |
| Diameter (English) Inner | 4.13 in. |
| Diameter (English) Outer | 5.51 in. |
| Diameter (Metric) Inner | 10.5 cm |
| Diameter (Metric) Outer | 14 cm |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| For Use With (Application) | Laboratory Applications Requiring a Variety of Temperature-Controlled Reactions and Useful in Biological and Chemical Reactions Including Titrations, Enzyme Digestions and Synthetic Organic Reactions |
| Height (English) Inner | 5.23 in. |
| Show More |