Learn More
DWK Life Sciences Kimble™ Borosilicate Micro Disposable Pipettes with Holder
Click to view available options
Length (Metric):
32 mm
41 mm
missing translation for 'unitSize'
Pack of 100
Description
- Inexpensive
- you can dispose of each tube after use
- Filling is simple
- Capillary action, not externally applied suction, draws the fluid into the tube
- To dispense the fluid, simply squeeze the bulb
Specifications
Specifications
| Certifications/Compliance | USP type I, ASTM E438 |
| Diameter (English) Inner | 0.0219 in. |
| Diameter (English) Outer | 0.0314 in. |
| Diameter (Metric) Inner | 0.5562 mm |
| Diameter (Metric) Outer | 0.7975 mm |
| Material | Borosilicate Glass |
| Includes | 1 Bulb assembly and 1 dispenser vial |
| Length (English) | 1.61 in. |
| Length (Metric) | 41 mm |
| Type | Micropipet |
| Show More |