Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ Reusable High Strength Centrifuge Tubes
Product Code. 11736923
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
Length (Metric):
102 mm
106 mm
Diameter (Metric):
18 mm
24 mm
Unit Size:
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Diameter (Metric) unitSize
11736923 30 mL 106 mm 24 mm Pack of 6
11844005 15 mL 102 mm 18 mm Pack of 6
11875824 30 mL 106 mm 24 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11736923 Supplier DWK Life Sciences Supplier No. 4560030
Chemically strengthened to achieve a greater mechanical strength
  • Made of KIMAX Type 1, Class B, borosilicate glass
  • Can be centrifuged up to 13,100xG with rubber adapter sleeve
  • Withstand temperatures up to 300°C
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM E438
Closure Material Phenolic
Capacity (Metric) 30 mL
Disposable Reusable
Material Borosilicate Glass
Liner Material PTFE, Rubber
Thread Size 24-400
Length (Metric) 106 mm
Capacity (English) 1.01 oz.
Diameter (Metric) 24 mm
Max. RCF 13,100 x g