Få mere at vide
Få mere at vide
DWK Life Sciences Kimble™ Kontes™ Hollow White Rubber Sleeve Stoppers
Klik for at se tilgængelige muligheder
Diameter (Metric):
1.4 cm
2.4 cm
Standard Taper Joint:
14/20
24/40; 24/20
Pakningsstørrelse:
Each
Pack of 50
Beskrivelse
Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.
- Plug-type stopper with sleeve extension fits into the neck of the apparatus for a secure seal
- Diaphragm can be punctured with a syringe needle
- Use for outgassing NMR sample tubes, as a pressure vent or for adding reactant via a syringe
Tekniske data
Tekniske data
| Diameter (Metric) | 2.4 cm |
| For Use With (Equipment) | NMR tubes, pressure venting or adding reactants via syringe |
| Standard Taper Joint | 24/40; 24/20 |
| Color | White |
| Material | Rubber |
| Product Type | Plug-type Sleeve Stopper |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion