Learn More
DWK Life Sciences Kimble™ Kontes™ Heavy Wall Recovery Flasks with Square Bead, standard taper Outer Joint
Product Code. 11892923
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
500 mL
1000 mL
Standard Taper Joint:
24/40
Includes:
Square Bead Joints
Unit Size:
Each
Product Code. 11892923
Brand: DWK Life Sciences 6086754624
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ideal for use with rotary evaporators
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
  • Single-neck flask with standard taper outer joint
  • Designed for easy recovery of reaction products
  • Total contents are accessible with a spatula or policeman
  • Lower portion fits standard heating mantles
  • Even wall thickness and minimum runout
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 4.01 in.
Capacity (Metric) 500 mL
Material Borosilicate Glass
For Use With (Application) Square bead joints provide superior clamping to standard Joint beads
For Use With (Equipment) Rotary Evaporator
Includes Square Bead Joints
Neck Style Single
No. per Case 1/Cs.
Standard Taper Joint 24/40
Diameter (Metric) Outer 102 mm