Learn More
DWK Life Sciences™ Kimble™ Kontes™ Flask Inlet Adapter, Monier-Williams, 24/40 Joint
Product Code. 15120526
Change view
Click to view available options
Standard Taper Joint:
24/40
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
15120526 24/40 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15120526 Supplier DWK Life Sciences™ Supplier No. 5138010000
  • ONLINE EXCLUSIVE PRICE
    112.35€
    118.84€ / Pack of 1
    Save POA
    -5.78% Off
 Restricted Item
Estimated Shipment: 08-04-2027
Component Parts of Monier-Williams Sulfites Distillation Apparatus which is used for the determination of free sulfite and a reproducible portion of the bound sulfites in food, such as the carbonyl addition products.
  • Flask Inlet Adapter
  • Standard Taper joint 24/40
  • Case of 1
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Type Flask Inlet Adapter
Standard Taper Joint 24/40