Learn More
Klever Kleen™ Kutter with Stainless Steel Blade
Descripción
- Recessed blade reduces cut injuries and damaged goods.
- Will cut boxes, film, tape and thousands more applications.
- Small, convient and durable with high quality carbon steel blade.
- Highest safety level.
- Disposable.
- Our antimicrobial Kleen™ safety cutters utilize an antimicrobial material that inhibits the growth of bacteria that may affect this product.
- The antimicrobial properties do not protect users or others against bacteria, viruses, germs, or other disease organisms.
- Clean and wash this product thoroughly per your industry's guidelines.
- The active antimicrobial component is part of the formulation, so it won't wash off or wear away.
- This is not a coating or treatment; it's an integral ingredient.
Especificaciones
Especificaciones
| Blade Material | Stainless Steel |
| Product Type | Kleen Cutter |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Color | Green |
| Quantity | 10/Pk. |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?