Learn More
Lenz Laborglasinstrumente™ Kjeldahl Flask
Product Code. 10529791
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10529791 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10529791 Supplier Lenz Laborglasinstrumente™ Supplier No. 03052958
  • ONLINE EXCLUSIVE PRICE
    808.00€
    549.17€ / Pack of 10
    Save 258.83€
    32% Off
Estimated Shipment: 16-12-2026
Specifications
Capacity (Metric) 500 mL
Includes With Conical Joint and Inscription Label
Diameter (Metric) Outer 101 mm