Learn More
Afora™ Kjeldahl Ball
Product Code. 12044822
Change view
Click to view available options
Material:
Classic Frosted Glass
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12044822 Classic Frosted Glass Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12044822 Supplier Afora™ Supplier No. 914/E
  • ONLINE EXCLUSIVE PRICE
    145.95€
    130.65€ / Pack of 1
    Save 15.30€
    10% Off
Estimated Shipment: 17-11-2026
Prevent boiling over that could result in distillate contamination with the Afora™ Kjeldahl Ball;
  • Weight: 0.01kg
  • 1Pack
Specifications
Description With Retention Trap
Material Classic Frosted Glass
Standard Taper Joint Male: 29/32; Female: 29/32