Learn More
Equip Labo™ Kit Lab™ Connection Kit
Product Code. 12556895
Change view
Click to view available options
Size: 200mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12556895 Size: 200mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12556895 Supplier Equip Labo™ Supplier No. 5512
  • ONLINE EXCLUSIVE PRICE
    149.00€
    123.84€ / Pack of 1
    Save 25.16€
    16% Off
 Restricted Item
Estimated Shipment: 08-01-2027