Learn More
Thermo Scientific™ KingFisher Plastic Consumables for SARS-CoV-2 Testing
Product Code. 17153249 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Duo Prime, Flex, Apex, Presto
KingFisher Flex 96
KingFisher Flex and Presto
Quantity:
100/Pk.
50/Pk.
Includes:
KF 96 DW plate, v-bottom
KingFisher DW 96 Tip Comb
Tip Comb Presenting Plate
Unit Size:
Pack of 100
Pack of 50
Product Code. 17153249
Brand: Thermo Scientific™ A48438
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
These KingFisher compatible plastic consumables are offered specifically for labs performing COVID-19 testing with the MagMAX Viral/Pathogen Nucleic Acid Purification Kit and the TaqPath COVID-19 Combo Kit.
Optimize performance using Thermo Scientific KingFisher Systems Consumables, designed specifically for use for COVID-19 testing with these instruments. KingFisher instruments utilize disposable plastics made of polypropylene. The plastics, tip combs and microplates, are ideal for magnetic particle processing because of their low binding affinity for biomolecules. The special design enables excellent recovery of magnetic beads.  Specifically tested with  the TaqPath COVID-19 Combo Kit and MagMAX Viral/Pathogen Nucleic Acid Isolation Kit.
For TaqPath COVID-19 testing with MagMAX Viral/Pathogen Nucleic Acid Isolation Kit
TRUSTED_SUSTAINABILITY
Specifications
Format 96-well Plate
Capacity 96 Samples
For Use With (Equipment) KingFisher Flex and Presto
Includes KingFisher DW 96 Tip Comb
Quantity 100/Pk.