Learn More
Thermo Scientific™ KingFisher Plastic Consumables for SARS-CoV-2 Testing
Shop All Thermo Scientific ProductsDescription
These KingFisher compatible plastic consumables are offered specifically for labs performing COVID-19 testing with the MagMAX Viral/Pathogen Nucleic Acid Purification Kit and the TaqPath COVID-19 Combo Kit.
For TaqPath COVID-19 testing with MagMAX Viral/Pathogen Nucleic Acid Isolation Kit
Specifications
Specifications
| Format | 96-well Plate |
| For Use With (Equipment) | KingFisher Flex 96 |
| Includes | Tip Comb Presenting Plate |
| Quantity | 50/Pk. |