Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 standard and PCR formats
Shop All Thermo Scientific ProductsDescription
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Specifications
Specifications
| Format | 96-well Comb |
| Capacity | 96 Samples |
| Description | KingFisher 96 tip comb for KingFisher magnets, 100 pcs (for Flex) |
| For Use With (Equipment) | KingFisher Flex |
| Quantity | 100/Cs. |