Learn More
Macherey-Nagel™ KingFisher™ Duo Prime Accessory Kit
Product Code. 15869167
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Product Code. 15869167
Brand: Macherey-Nagel™ 744952
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Enables automated purification of genomic DNA using a NucleoMag™ kit on the KingFisher™ Duo / Duo Prime platform for faster, high throughput performance with reliable yields.
  • Adapted NucleoMag™ procedure for instruments of the KingFisher™ Duo/Duo Prime series
  • Reliable performance and excellent yields from different sample materials
  • Speeds up nucleic acid purification workflow: process 96 samples in less than 30 minutes (excluding lysis)
  • Set includes Deep-well Plates, Tip Combs and Elution Strips for processing in 96-well format
Specifications
For Use With (Equipment) KingFisher™ Duo/Duo Prime platform, NucleoMag™ kit
For Use With (Application) Automated nucleic acid purification
Includes Deep-well Plates, Tip Combs and Elution Strips for processing in 96-well format
Quantity 1 Set