Learn More
Thermo Scientific™ KingFisher Duo Prime Accessories
Product Code. 30132313 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
24 Deep-well Head
24 Deep-well Heating Block
96 Deep-well Head
96 Deep-well Heating Block
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
30132313 24 Deep-well Head Each
30131189 96 Deep-well Head Each
30131520 24 Deep-well Heating Block Each
30131332 96 Deep-well Heating Block Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30132313 Supplier Thermo Scientific™ Supplier No. N12460
 Restricted Item
Estimated Shipment: 15-12-2026
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources.
The KingFisher Duo Prime system is a flexible system for sample processing that offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, and cell purification from a wide range of sources. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality.
The KingFisher Duo Prime instrument comes with a 12-pin magnet head for 96 deep well applications, a 6-pin magnet head for 24 deep well applications, and heating blocks. Additional magnet heads and heating blocks are offered here for separate purchase.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Nucleic Acid Extraction and Cell Separation Instrument
Sample Type Fresh or Frozen Plant Tissues, FFPE Tissue, Blood, Cells, Plasma
Volume (Metric) Processing 200 to 5000 μL
Format 24 Deep-well Head
Capacity 6 Samples
Disposable Reusable
For Use With (Equipment) KingFisher Duo Prime Purification System
Quantity 1 Head