Learn More
Thermo Scientific™ KingFisher™ Apex Dx Magnetic Heads
Product Code. p-200000116 Shop All Thermo Scientific Products
Click to view available options
Format:
Apex Dx 24 Combi Head
Apex Dx 96 Combi Head
Apex Dx 96 Deep Well Head
Apex Dx 96 PCR Head
Unit Size:
Each
Product Code. 17846588
Brand: Thermo Scientific™ B24079940
The Thermo Scientific KingFisher Apex Dx Purification Instrument allows for extraction of nucleic acids from human specimens with a small, compact footprint making it ideal for use in diagnostic and development workflows.
KingFisher Apex Dx Magnetic Heads are for use with the KingFisher Apex Dx Purification Instrument. The instrument accommodates two interchangeable magnetic heads (it comes with one included). The interchangeable heads and heating blocks provide flexibility in the extraction of nucleic acids, proteins, and cells with a small, compact footprint, making the instrument ideal for use in diagnostic and development workflows.
With its intuitive software, automation capabilities, and revolutionary magnetic particle separation technology, the KingFisher Apex Dx Purification Instrument provides high quality, exceptional reliability, and outstanding run-to-run reproducibility for clinicians and assay developers seeking an IVD sample preparation solution for their diagnostic workflow.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Nucleic Acid Extraction and Cell Separation Instrument
Sample Type Viral Samples
Format Apex Dx 24 Combi Head
Disposable Reusable
Quantity 1 head
For in vitro diagnostic use.