Learn More
Thermo Scientific™ KingFisher Apex Purification Systems
Product Code. p-8929004 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
24 Combi head
96 Combi head
96 DW head
96 PCR Head
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
16592442 96 PCR Head Each
16502452 96 Combi head Each
16512452 96 DW head Each
16522452 24 Combi head Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16592442 Supplier Thermo Scientific™ Supplier No. 5400910
This item requires a price update prior to being ordered.
Request a Price for this item here.
Currently Unavailable: We have no information when this product will be available.
View alternative products
The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source.
The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. It moves magnetic beads, not liquids, through the main steps of sample preparation: bind, wash, and elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing.
  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
TRUSTED_SUSTAINABILITY
Specifications
Product Type 96 PCR Head
Sample Type Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate
Suitable Downstream Analysis PCR/qPCR, Sequencing, Mass Spec
Volume (Metric) Processing 10 to 80 μL
Plates per Deck 8
Capacity 96 Samples
Collection Efficiency of Particles >98% with 2.8 μm particle size
Cooling Temperature Down to +40°C
Display Touch screen (10 in.)
For Use With (Application) Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating Temperature +4°C above ambient temperature up to +100°C
Magnetic Rods 96
Particle Size (Metric) ca. >1 μm
Samples per Run 96
Type of Magnet Head 96 in one frame, interchangeable
UV Exposure Time Up to 23 hr. 59 min.
UV Lamp 2 pcs
Plate Type 96 PCR plate (semi/skirted)
Depth (English) 22 in.
Depth (Metric) 56 cm
Height (English) 18 in.
Height (Metric) 46 cm
Weight (English) 123 lb.
Weight (Metric) 56 kg
Width (English) 31 in.
Width (Metric) 79 cm
Show More Show Less
Can I use the KingFisher Flex Purfication System standard magnet with the new KingFisher Apex Purification System?
Unfortunately, the heads are not interchangeable between KingFisher Flex Purification System and KingFisher Apex Purification System.
How many kinds (type or size) of magnets are there for the KingFisher Apex 96 DW Head (Cat. No. 24079930)?
Please see the Thermo Scientific KingFisher Flex User Manual, page 32. As you will see, there are four different heads. In terms of magnet force, it's a permanent magnet so the expected life is >100 years.
What should I do if the tip comb does not load correctly onto my Thermo Scientific KingFisher system?
If the tip does not load correctly onto a Thermo Scientific KingFisher magnetic head, we recommend that you try manually flexing the tips both lengthwise and widthwise to level the tip comb. A video of flexing the tip comb can be found here: https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/automated-purification-extraction/kingfisher-systems/resources.html. The video is under "KingFisher Flex" and titled "How to install the magnet head and check the tip comb." Flexing of the tip comb is demonstrated at timepoint 1:35. If that does not work, try a different box or lot of tips. This applies to all KingFisher instruments.