Learn More
HMC Europe Vertical Autoclaves: HV Series
Programmable top-loading autoclaves with varying chamber volumes
Brand: HMC Europe 20003
Description
- Use steam sterilization
- Six sizes from 25 to 110 liters
- Low height, easy-to-load
- Portable — require no plumbing to operate but can be plumbed
- Complete line of baskets, inserts and cans available separately
- HV Series (dependable, hard-working, economical)
- Many upgrades available depending on your needs
Certifications
CE marked
Prestations de service
Two years on parts; One year on labor
Spécification
CE | |
100 cm | |
42 cm | |
110 L | |
31.1 in. | |
HV L110 | |
105°C to 123°C | |
81 kg | |
25.59 in. | |
25.98 in. |
39.37 in. | |
16.53 in. | |
29 gal. | |
Steam sterilization | |
79 cm | |
221°F to 253.4°F | |
178.57 lb. | |
65 cm | |
4 w | |
66 cm |